Assay Details
Target Gene Details
Entrez Gene ID: | 12759 |
Gene Name: | clusterin |
Gene Aliases: |
AI893575, ApoJ, Cli, D14Ucla3, SP-40, Sgp-2, Sgp2, Sugp-2 |
Location: |
Chr.14:65968483-65981547 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clu | NM_013492.2 | NP_038520.2 | ||
| AF248057.1 | AAF67184.1 | |||
| AK132030.1 | BAE20954.1 | |||
| BC075668.1 | AAH75668.1 | |||
| BY248781.1 | ||||
| D14077.1 | BAA03162.1 | |||
| L05670.1 | AAA37284.1 | |||
| L08235.1 | AAA37422.1 | |||
| S70244.1 | AAB30623.1 |

Genomic Map