Assay Details
Target Gene Details
Entrez Gene ID: | 20707 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 9c |
Gene Aliases: |
3830421J05Rik, Nk9, Spi11 |
Location: |
Chr.13:33149275-33159754 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb9c | NM_001164524.1 | 6 | 729 | NP_001157996.1 |
| NM_011453.6 | 6 | 700 | NP_035583.1 | |
| AK014448.1 | 6 | 704 | BAB29358.1 | |
| AK017139.1 | 6 | 718 | ||
| BC051664.1 | 6 | 700 | AAH51664.1 |
More Information
Set Membership: |
|

Genomic Map