Assay Details
Target Gene Details
Entrez Gene ID: | 110596 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 28 |
Gene Aliases: |
9230110L08Rik, AI323540, D13Bwg1089e, RIP2, Rgnef, RhoGEF, Rhoip2, p190RhoGEF |
Location: |
Chr.13:97898594-98206624 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef28 | NM_012026.2 | 27 | 3573 | NP_036156.2 |
| AK029167.1 | 21 | 2733 | ||
| AK048676.1 | 19 | 2539 | ||
| AK173325.1 | 27 | 3596 | BAD32603.1 | |
| BC138030.1 | 27 | 3497 | ||
| U73199.1 | 27 | 3573 | AAB18197.1 |

Genomic Map