Assay Details
Target Gene Details
Entrez Gene ID: | 102216272 |
Gene Name: | adenylate kinase 6 |
Gene Aliases: |
2810046E22Rik, 4921516M08Rik, AK/ATPase |
Location: |
Chr.13:100651343-100666415 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak6 | NM_027592.3 | NP_081868.1 | ||
| AK012905.1 | ||||
| AK014903.1 | ||||
| BC019453.1 | AAH19453.1 |
Target Gene Details
Entrez Gene ID: | 108143 |
Gene Name: | TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
2310012M09Rik, AA673500, AU045423, TAFII31, TAFII32, Taf2g |
Location: |
Chr.13:100651343-100656060 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf9 | NM_001015889.2 | NP_001015889.1 | ||
| NM_027139.5 | NP_081415.1 | |||
| AF305839.1 | AAL47693.1 | |||
| BC042723.1 | AAH42723.1 | |||
| BC108348.1 | AAI08349.1 | |||
| BQ839921.1 | ||||
| BU946135.1 | ||||
| BY108476.1 |
More Information
Set Membership: |
|

Genomic Map