Assay Details
Target Gene Details
Entrez Gene ID: | 102216272 |
Gene Name: | adenylate kinase 6 |
Gene Aliases: |
2810046E22Rik, 4921516M08Rik, AK/ATPase |
Location: |
Chr.13:100651343-100666415 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak6 | NM_027592.3 | NP_081868.1 | ||
| AK012905.1 | ||||
| AK014903.1 | ||||
| BC019453.1 | AAH19453.1 | |||
| BG084768.2 |
More Information
Set Membership: |
|

Genomic Map