Assay Details
Target Gene Details
Entrez Gene ID: | 218343 |
Gene Name: | tetratricopeptide repeat domain 37 |
Gene Aliases: |
mKIAA0372 |
Location: |
Chr.13:76098730-76190323 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc37 | NM_001081352.1 | NP_001074821.1 | ||
| AK134255.1 | BAE22066.1 | |||
| AK164627.1 |

Genomic Map