Assay Details
Target Gene Details
Entrez Gene ID: | 218343 |
Gene Name: | tetratricopeptide repeat domain 37 |
Gene Aliases: |
mKIAA0372 |
Location: |
Chr.13:76098730-76190323 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc37 | NM_001081352.1 | NP_001074821.1 | ||
| AK129128.1 | BAC97938.1 | |||
| AK162476.1 |

Genomic Map