Assay Details
Target Gene Details
Entrez Gene ID: | 74338 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19 |
Gene Aliases: |
4632401C08Rik, B<0>AT1 |
Location: |
Chr.13:73679745-73700695 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a19 | NM_028878.3 | NP_083154.1 | ||
| AB096860.1 | BAC66209.1 | |||
| AJ633679.1 | CAG17898.1 | |||
| AK014544.1 | BAB29422.1 | |||
| AK028784.1 | BAC26119.1 | |||
| AK128923.1 | ||||
| AK162399.1 | BAE36894.1 | |||
| BC107411.1 | AAI07412.1 | |||
| BC107412.2 | AAI07413.1 |
More Information
Set Membership: |
|

Genomic Map