Assay Details
Target Gene Details
Entrez Gene ID: | 104416 |
Gene Name: | Brca1 associated protein 1 |
Gene Aliases: |
2300006C11Rik, AA989761, AW553466, mKIAA0272, uch-x4 |
Location: |
Chr.14:31251455-31259930 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bap1 | NM_027088.2 | NP_081364.1 | ||
| AB047820.1 | BAB32976.1 | |||
| AK009033.1 | ||||
| AK079374.1 | ||||
| AK129108.1 | BAC97918.1 | |||
| AK170576.1 | BAE41889.1 | |||
| AK171561.1 | ||||
| AK181980.1 | ||||
| BC050901.1 | AAH50901.1 |
More Information
Set Membership: |
|

Genomic Map