Assay Details
Target Gene Details
Entrez Gene ID: | 104416 |
Gene Name: | Brca1 associated protein 1 |
Gene Aliases: |
2300006C11Rik, AA989761, AW553466, mKIAA0272, uch-x4 |
Location: |
Chr.14:31251455-31259930 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bap1 | NM_027088.2 | 5 | 448 | NP_081364.1 |
| AB047820.1 | 5 | 349 | BAB32976.1 | |
| AK079374.1 | 5 | 464 | ||
| AK129108.1 | 5 | 356 | BAC97918.1 | |
| AK170576.1 | 5 | 449 | BAE41889.1 | |
| AK171561.1 | 5 | 450 | ||
| AK178991.1 | 2 | 114 | ||
| AK183536.1 | 2 | 114 | ||
| AK185015.1 | 5 | 328 | ||
| AK198656.1 | 2 | 114 | ||
| AK217563.1 | 2 | 114 | ||
| BC050901.1 | 5 | 411 | AAH50901.1 |
More Information
Set Membership: |
|

Genomic Map