Assay Details
Target Gene Details
Entrez Gene ID: | 21810 |
Gene Name: | transforming growth factor, beta induced |
Gene Aliases: |
68kDa, AI181842, AI747162, Beta-ig, big-h3 |
Location: |
Chr.13:56609603-56639339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfbi | NM_009369.4 | NP_033395.1 | ||
| AK084431.1 | BAC39181.1 | |||
| AK135536.1 | BAE22571.1 | |||
| AK142323.1 | BAE25032.1 | |||
| AK150449.1 | ||||
| AK150833.1 | ||||
| AK150969.1 | BAE30000.1 | |||
| AK151680.1 | BAE30605.1 | |||
| AK152365.1 | BAE31155.1 | |||
| AK152662.1 | ||||
| AK155572.1 | BAE33330.1 | |||
| AK155828.1 | BAE33452.1 | |||
| AK159711.1 | ||||
| AK164373.1 | ||||
| AK169912.1 | BAE41453.1 | |||
| AK170495.1 | BAE41834.1 | |||
| AK171557.1 | BAE42525.1 | |||
| BC129900.1 | ||||
| BC129901.1 | ||||
| L19932.1 | AAC37658.1 |
More Information
Set Membership: |
|

Genomic Map