Assay Details
Target Gene Details
Entrez Gene ID: | 27015 |
Gene Name: | polymerase (DNA directed), kappa |
Gene Aliases: |
Dinb1 |
Location: |
Chr.13:96480689-96542648 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polk | NM_012048.2 | NP_036178.1 | ||
| AB027563.1 | BAA86942.1 | |||
| AF163571.1 | AAF02541.1 | |||
| AY769919.1 | AAV80817.1 | |||
| AY769920.1 | AAV80818.1 | |||
| AY769921.1 | AAV80819.1 | |||
| AY769922.1 | AAV80820.1 | |||
| AY769923.1 | AAV80821.1 | |||
| AY769924.1 | AAV80822.1 | |||
| AY769925.1 | AAV80823.1 | |||
| AY769926.1 | AAV80824.1 | |||
| AY769927.1 | AAV80825.1 | |||
| AY769928.1 | AAV80826.1 | |||
| BC052820.1 | AAH52820.1 |
More Information
Set Membership: |
|

Genomic Map