Assay Details
Target Gene Details
Entrez Gene ID: | 19210 |
Gene Name: | phosphatidylserine synthase 1 |
Gene Aliases: |
AU044268, AW539008, PSS-1, mKIAA0024 |
Location: |
Chr.13:66932830-66998401 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptdss1 | NM_008959.3 | NP_032985.2 | ||
| AF042731.1 | AAB97845.1 | |||
| AK036990.1 | BAC29660.1 | |||
| AK054101.1 | BAC35656.1 | |||
| AK085857.1 | BAC39555.1 | |||
| AK086136.1 | ||||
| AK088048.1 | BAC40117.1 | |||
| AK137681.1 | BAE23459.1 | |||
| AK172873.1 | BAD32151.1 | |||
| AK219321.1 | ||||
| BC003260.1 | AAH03260.1 |
More Information
Set Membership: |
|

Genomic Map