Assay Details
Target Gene Details
Entrez Gene ID: | 20499 |
Gene Name: | solute carrier family 12, member 7 |
Gene Aliases: |
AA408796, D13Ertd261e, Kcc4 |
Location: |
Chr.13:73733053-73816755 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a7 | NM_011390.2 | NP_035520.1 | ||
| AF087436.1 | AAD38328.1 | |||
| AK131129.1 | BAD21379.1 | |||
| AK143535.1 | ||||
| AK149750.1 | BAE29061.1 | |||
| AK166215.1 | ||||
| AK169950.1 | BAE41477.1 | |||
| AK171498.1 | BAE42491.1 | |||
| BC141107.1 |
More Information
Set Membership: |
|

Genomic Map