Assay Details
Target Gene Details
Entrez Gene ID: | 13522 |
Gene Name: | a disintegrin and metallopeptidase domain 28 |
Gene Aliases: |
C130072N01Rik, D430033C21Rik, Dtgn1, MDC-L, MDC-Lm, MDC-Ls, MDCL, TECADAM, eMDCII |
Location: |
Chr.14:68604994-68655887 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam28 | NM_001048175.2 | NP_001041640.1 | ||
| NM_010082.2 | NP_034212.1 | |||
| NM_183366.3 | NP_899222.1 | |||
| NR_102399.1 | ||||
| AF153350.1 | AAF71993.1 | |||
| AF163290.1 | AAM21935.1 | |||
| AF163291.1 | AAM21936.1 | |||
| AF163292.1 | AAM21937.1 | |||
| AF163293.1 | AAQ07988.1 | |||
| AK136715.1 | ||||
| BC058782.1 | AAH58782.1 |
More Information
Set Membership: |
|

Genomic Map