Assay Details
Target Gene Details
Entrez Gene ID: | 12050 |
Gene Name: | BCL2-like 2 |
Gene Aliases: |
AW048834, Bcl-w, Gtrgal2, Gtrosa41, bclw, c98 |
Location: |
Chr.14:54883366-54889095 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l2 | NM_007537.1 | NP_031563.1 | ||
| AF030769.1 | AAB86430.1 | |||
| AK004680.1 | BAB23468.1 | |||
| AK015644.1 | BAB29912.1 | |||
| AY170344.3 | AAO13177.2 |

Genomic Map