Assay Details
Target Gene Details
Entrez Gene ID: | 20499 |
Gene Name: | solute carrier family 12, member 7 |
Gene Aliases: |
AA408796, D13Ertd261e, Kcc4 |
Location: |
Chr.13:73733053-73816755 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a7 | NM_011390.2 | 4 | 477 | NP_035520.1 |
| AF087436.1 | 4 | 479 | AAD38328.1 | |
| AK052910.1 | 4 | 528 | BAC35198.1 | |
| AK083598.1 | 3 | 865 | ||
| AK131129.1 | 4 | 431 | BAD21379.1 | |
| AK143535.1 | 4 | 471 | ||
| AK149750.1 | 4 | 472 | BAE29061.1 | |
| AK166215.1 | 4 | 471 | ||
| AK169950.1 | 4 | 467 | BAE41477.1 | |
| AK171498.1 | 4 | 514 | BAE42491.1 | |
| BC141107.1 | 4 | 437 |
More Information
Set Membership: |
|

Genomic Map