Assay Details
Target Gene Details
Entrez Gene ID: | 11495 |
Gene Name: | a disintegrin and metallopeptidase domain 2 |
Gene Aliases: |
AI323749, Ftnb, Ph30-beta |
Location: |
Chr.14:66027329-66077856 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam2 | NM_009618.2 | 16 | 1727 | NP_033748.2 |
| AK016550.1 | 16 | 1727 | BAB30298.1 | |
| AK161266.1 | 16 | 1716 | BAE36279.1 | |
| U16242.1 | 16 | 1716 | AAA90980.1 | |
| U22057.1 | 15 | 1667 | AAA74921.1 | |
| U38806.1 | 16 | 1732 | AAD04207.1 |
More Information
Set Membership: |
|

Genomic Map