Assay Details
Target Gene Details
Entrez Gene ID: | 11495 |
Gene Name: | a disintegrin and metallopeptidase domain 2 |
Gene Aliases: |
AI323749, Ftnb, Ph30-beta |
Location: |
Chr.14:66027329-66077856 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam2 | NM_009618.2 | NP_033748.2 | ||
| AK016550.1 | BAB30298.1 | |||
| AK161261.1 | ||||
| AK161266.1 | BAE36279.1 | |||
| BY079316.1 | ||||
| U16242.1 | AAA90980.1 | |||
| U22057.1 | AAA74921.1 | |||
| U38806.1 | AAD04207.1 |
More Information
Set Membership: |
|

Genomic Map