Assay Details
Target Gene Details
Entrez Gene ID: | 19012 |
Gene Name: | phosphatidic acid phosphatase type 2A |
Gene Aliases: |
Hic53, Hpic53, LPP-1, LPP1, PAP-2a, PAP2-alpha, PAP2a, mPAP |
Location: |
Chr.13:112800727-112867894 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppap2a | NM_008247.3 | 6 | 1234 | NP_032273.1 |
| NM_008903.2 | 6 | 1231 | NP_032929.1 | |
| AK077275.1 | 6 | 1118 | BAC36724.1 | |
| AK190923.1 | ||||
| AY247795.1 | 6 | 856 | AAP04434.1 | |
| AY247796.1 | 6 | 859 | AAP04435.1 | |
| BC061161.1 | 6 | 1219 | AAH61161.1 | |
| CD742481.1 | 1 | 396 | ||
| D84376.1 | 6 | 1085 | BAA12335.1 | |
| L43371.1 | 6 | 1144 | AAA85353.1 |
More Information
Set Membership: |
|

Genomic Map