Assay Details
Target Gene Details
Entrez Gene ID: | 105239 |
Gene Name: | ring finger protein 44 |
Gene Aliases: |
AI854545, mKIAA1100 |
Location: |
Chr.13:54679399-54693995 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf44 | NM_001146025.1 | NP_001139497.1 | ||
| NM_001146026.1 | NP_001139498.1 | |||
| NM_001146027.1 | NP_001139499.1 | |||
| NM_134064.2 | NP_598825.2 | |||
| NR_027395.1 | ||||
| NR_027396.1 | ||||
| AK147349.1 | BAE27859.1 | |||
| AK160090.1 | BAE35621.1 | |||
| AK160115.1 | BAE35639.1 | |||
| AK161651.1 | ||||
| AK171943.1 | BAE42740.1 | |||
| AK172257.1 | ||||
| BC017630.1 | AAH17630.1 | |||
| BC035548.1 | AAH35548.1 | |||
| CF728349.1 | ||||
| CJ147978.1 |

Genomic Map