Assay Details
Target Gene Details
Entrez Gene ID: | 20354 |
Gene Name: | sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D |
Gene Aliases: |
CD100, Semacl2, Semaj, Semcl2, coll-4 |
Location: |
Chr.13:51685522-51793747 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema4d | NM_001281880.1 | 15 | 2135 | NP_001268809.1 |
| NM_013660.4 | 14 | 2071 | NP_038688.2 | |
| AK088653.1 | 11 | 1353 | BAC40480.1 | |
| AK132688.1 | 15 | 2061 | BAE21302.1 | |
| AK150911.1 | 13 | 1963 | ||
| BC049780.1 | 14 | 1968 | AAH49780.2 | |
| U69535.1 | 12 | 1598 | AAC52964.1 |
More Information
Set Membership: |
|

Genomic Map