Assay Details
Target Gene Details
Entrez Gene ID: | 11500 |
Gene Name: | a disintegrin and metallopeptidase domain 7 |
Gene Aliases: |
9230104J12Rik, EAP1, EAPI |
Location: |
Chr.14:68497336-68533689 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam7 | NM_007402.2 | 13 | 1318 | NP_031428.2 |
| AF013107.1 | 13 | 1304 | AAC53368.1 | |
| AK136692.1 | 13 | 1317 | ||
| KF144368.1 | 1 | 41 | ||
| KF144369.1 | 1 | 41 |
More Information
Set Membership: |
|

Genomic Map