Assay Details
Target Gene Details
Entrez Gene ID: | 27062 |
Gene Name: | Ca2+-dependent secretion activator |
Gene Aliases: |
AU067781, CAPS, CAPS1, mKIAA1121 |
Location: |
Chr.14:12372560-12823314 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps | NM_001042617.1 | 8 | 1848 | NP_001036082.1 |
| NM_012061.3 | 8 | 1848 | NP_036191.2 | |
| AK122453.1 | 8 | 1848 | BAC65735.1 | |
| CB245630.1 | 3 | 300 | ||
| CF539197.1 | 5 | 568 | ||
| D86214.1 | 8 | 1891 | BAA13044.1 |
More Information
Set Membership: |
|

Genomic Map