Assay Details
Target Gene Details
Entrez Gene ID: | 27062 |
Gene Name: | Ca2+-dependent secretion activator |
Gene Aliases: |
AU067781, CAPS, CAPS1, mKIAA1121 |
Location: |
Chr.14:12372560-12823314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps | NM_001042617.1 | NP_001036082.1 | ||
| NM_012061.3 | NP_036191.2 | |||
| AK122453.1 | BAC65735.1 | |||
| AK133310.1 | ||||
| CB245630.1 | ||||
| CF539197.1 | ||||
| D86214.1 | BAA13044.1 |
More Information
Set Membership: |
|

Genomic Map