Assay Details
Target Gene Details
Entrez Gene ID: | 218442 |
Gene Name: | serine incorporator 5 |
Gene Aliases: |
A130038L21Rik, AIGP3, C86123, TPO1 |
Location: |
Chr.13:92611138-92711947 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serinc5 | NM_172588.2 | 2 | 234 | NP_766176.1 |
| AB029501.1 | 2 | 123 | BAC44829.1 | |
| AK037699.1 | 2 | 234 | BAC29851.1 | |
| AK054475.1 | 2 | 245 | BAC35794.1 | |
| AK136799.1 | 2 | 282 | BAE23132.1 | |
| AK163675.1 | 2 | 199 | ||
| AK165334.1 | 2 | 241 | BAE38139.1 | |
| BC049189.1 | 2 | 234 | AAH49189.2 | |
| BC062131.1 | 2 | 234 | AAH62131.1 |
More Information
Set Membership: |
|

Genomic Map