Assay Details
Target Gene Details
Entrez Gene ID: | 218442 |
Gene Name: | serine incorporator 5 |
Gene Aliases: |
A130038L21Rik, AIGP3, C86123, TPO1 |
Location: |
Chr.13:92611138-92711947 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serinc5 | NM_172588.2 | NP_766176.1 | ||
| AB029501.1 | BAC44829.1 | |||
| AK037699.1 | BAC29851.1 | |||
| AK054475.1 | BAC35794.1 | |||
| AK136799.1 | BAE23132.1 | |||
| AK165334.1 | BAE38139.1 | |||
| BC049189.1 | AAH49189.2 | |||
| BC062131.1 | AAH62131.1 |
More Information
Set Membership: |
|

Genomic Map