Assay Details
Target Gene Details
Entrez Gene ID: | 11500 |
Gene Name: | a disintegrin and metallopeptidase domain 7 |
Gene Aliases: |
9230104J12Rik, EAP1, EAPI |
Location: |
Chr.14:68497336-68533689 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam7 | NM_007402.2 | NP_031428.2 | ||
| AF013107.1 | AAC53368.1 | |||
| AK136692.1 | ||||
| AK164965.1 | BAE37982.1 |
More Information
Set Membership: |
|

Genomic Map