Assay Details
Target Gene Details
Entrez Gene ID: | 58860 |
Gene Name: | ADAM-like, decysin 1 |
Gene Aliases: |
2210414L24Rik, AI574231, Dcsn |
Location: |
Chr.14:68563387-68582072 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamdec1 | NM_021475.2 | 1 | 59 | NP_067450.1 |
| AJ242912.1 | 1 | 59 | CAB54557.1 | |
| AK156586.1 | 1 | 74 | BAE33767.1 | |
| BC046324.1 | AAH46324.1 |
More Information
Set Membership: |
|

Genomic Map