Assay Details
Target Gene Details
Entrez Gene ID: | 66082 |
Gene Name: | abhydrolase domain containing 6 |
Gene Aliases: |
0610041D24Rik, AA673485, AV065425 |
Location: |
Chr.14:8002807-8056773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd6 | NM_025341.3 | NP_079617.2 | ||
| AK002883.1 | BAB22430.1 | |||
| AK011887.1 | ||||
| AK018260.1 | BAB31136.1 | |||
| AK076105.1 | BAC36186.1 | |||
| AK090076.1 | BAC41081.1 | |||
| AK168782.1 | BAE40616.1 | |||
| BC027011.1 | AAH27011.1 |
More Information
Set Membership: |
|

Genomic Map