Assay Details
Target Gene Details
Entrez Gene ID: | 68750 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
1110037N09Rik, AA414966, B930013M22Rik |
Location: |
Chr.13:37778382-37952005 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rreb1 | NM_001039188.1 | NP_001034277.1 | ||
| NM_001177868.1 | NP_001171339.1 | |||
| NM_001177869.1 | NP_001171340.1 | |||
| NM_026830.2 | NP_081106.1 | |||
| NR_033218.1 | ||||
| NR_033615.1 | ||||
| AK047047.1 | ||||
| AK047251.1 | ||||
| AK147260.1 | BAE27803.1 | |||
| AK147653.1 | BAE28051.1 | |||
| AK154980.1 | BAE32969.1 | |||
| AK170652.1 | ||||
| AK171375.1 | BAE42417.1 | |||
| BC080680.1 | AAH80680.1 | |||
| BY202534.1 | ||||
| BY205187.1 | ||||
| BY206933.1 |

Genomic Map