Assay Details
Target Gene Details
Entrez Gene ID: | 66970 |
Gene Name: | single-stranded DNA binding protein 2 |
Gene Aliases: |
1500004K09Rik, 2310079I02Rik, 9330163K02Rik, A830008M03Rik, AU067692, AW558684, Hspc116, SSDP2 |
Location: |
Chr.13:91460979-91706175 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssbp2 | NM_024186.5 | NP_077148.1 | ||
| NM_024272.4 | NP_077234.2 | |||
| AK010220.1 | ||||
| AK013177.1 | BAB28693.1 | |||
| AK051840.1 | BAC34786.1 | |||
| AK133699.1 | ||||
| AY037837.1 | AAK71881.1 | |||
| AY167988.1 | AAN87334.1 | |||
| BC002176.1 | AAH02176.1 | |||
| EF410127.1 |
More Information
Set Membership: |
|

Genomic Map