Assay Details
Target Gene Details
Entrez Gene ID: | 238799 |
Gene Name: | transportin 1 |
Gene Aliases: |
AU021749, D13Ertd688e, IPO2, Kpnb2, MIP, MIP1, TRN |
Location: |
Chr.13:98839019-98926384 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnpo1 | NM_001048267.1 | NP_001041732.1 | ||
| NM_178716.3 | NP_848831.2 | |||
| AK029953.1 | BAC26696.1 | |||
| AK030580.1 | BAC27029.1 | |||
| AK037001.1 | ||||
| AK045132.1 | BAC32236.1 | |||
| AK167049.1 | BAE39215.1 | |||
| BC055372.1 | AAH55372.1 |
More Information
Set Membership: |
|

Genomic Map