Assay Details
Target Gene Details
Entrez Gene ID: | 97848 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6c |
Gene Aliases: |
C76171, C76174, SPI3C, SPIC, ovalbumin |
Location: |
Chr.13:33879816-33905708 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb6c | NM_148942.2 | NP_683744.2 | ||
| AF425084.1 | AAL65910.1 | |||
| BC062169.1 | AAH62169.1 |
More Information
Set Membership: |
|

Genomic Map