Assay Details
Target Gene Details
Entrez Gene ID: | 13135 |
Gene Name: | defender against cell death 1 |
Gene Aliases: |
AI323713 |
Location: |
Chr.14:54235485-54253929 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dad1 | NM_001113358.1 | NP_001106829.1 | ||
| NM_010015.4 | NP_034145.1 | |||
| AK002446.1 | BAB22107.1 | |||
| AK006711.1 | BAB24711.1 | |||
| AK030925.1 | BAE20464.1 | |||
| AK054480.1 | BAC35796.1 | |||
| BC024378.1 | AAH24378.1 | |||
| BC053379.1 | AAH53379.1 | |||
| BC058116.1 | AAH58116.1 | |||
| U22107.1 | AAA85855.1 | |||
| U83628.1 | AAC53359.1 |
More Information
Set Membership: |
|

Genomic Map