Assay Details
Target Gene Details
Entrez Gene ID: | 54616 |
Gene Name: | exostoses (multiple)-like 3 |
Gene Aliases: |
2900009G18Rik, AU040105, Ext1l, mKIAA0519 |
Location: |
Chr.14:65052059-65149899 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Extl3 | NM_018788.3 | 4 | 2872 | NP_061258.2 |
| AF083550.1 | 2 | 2662 | AAD42040.1 | |
| AK137842.1 | 2 | 1949 | BAE23506.1 | |
| AK220344.1 | 2 | 2215 | BAD90409.1 | |
| BC065073.1 | 2 | 2337 | AAH65073.1 |
More Information
Set Membership: |
|

Genomic Map