Assay Details
Target Gene Details
Entrez Gene ID: | 74195 |
Gene Name: | elongator acetyltransferase complex subunit 3 |
Gene Aliases: |
2610507P14Rik, KAT9 |
Location: |
Chr.14:65530446-65593112 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elp3 | NM_001253812.1 | 10 | 1131 | NP_001240741.1 |
| NM_028811.3 | 10 | 1074 | NP_083087.1 | |
| NR_045599.1 | 10 | 1074 | ||
| AK012072.1 | 10 | 1012 | BAB28009.1 | |
| AK088457.1 | 10 | 1095 | BAC40364.1 | |
| AK150058.1 | 10 | 1012 | ||
| AK208934.1 | 2 | 90 | ||
| AK209392.1 | 2 | 90 | ||
| BC026461.1 | 8 | 726 | AAH26461.1 | |
| BC057453.1 | 10 | 1014 | AAH57453.1 |
More Information
Set Membership: |
|

Genomic Map