Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmpr2 | NM_177992.2 | NP_818773.1 | ||
| AK017821.1 | ||||
| AK031004.1 | BAC27211.1 | |||
| AK075772.1 | BAC35946.1 | |||
| AK163048.1 | 1 | 102 | ||
| BC003886.1 | AAH03886.1 | |||
| BC024109.1 | AAH24109.1 | |||
| BY099107.1 |
More Information
Set Membership: |
|

Genomic Map