Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmpr2 | NM_177992.2 | NP_818773.1 | ||
| AK031004.1 | BAC27211.1 | |||
| AK075772.1 | BAC35946.1 | |||
| AK179292.1 | ||||
| BC003886.1 | AAH03886.1 | |||
| BC024109.1 | AAH24109.1 |
Target Gene Details
Entrez Gene ID: | 28113 |
Gene Name: | Terf1 (TRF1)-interacting nuclear factor 2 |
Gene Aliases: |
AW552114, D14Wsu146e, Tin2 |
Location: |
Chr.14:55676208-55682624 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tinf2 | AK028952.1 | 8 | 2907 | |
| AK144592.1 | BAE25954.1 | |||
| AK169480.1 | 3 | 2899 |
More Information
Set Membership: |
|

Genomic Map