Assay Details
Target Gene Details
Entrez Gene ID: | 239188 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 1 |
Gene Aliases: |
B230207J08, D230005D02Rik |
Location: |
Chr.14:77154001-77721763 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enox1 | NM_001253759.1 | NP_001240688.1 | ||
| NM_172813.3 | NP_766401.1 | |||
| AK045500.1 | ||||
| AK051821.1 | BAC34782.1 | |||
| AK082388.1 | ||||
| AK162001.1 | ||||
| BC052062.1 | AAH52062.1 |
Target Gene Details
Entrez Gene ID: | 629678 |
Gene Name: | predicted gene 6994 |
Gene Aliases: |
EG629678 |
Location: |
Chr.14:77479483-77506868 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6994 | NR_033141.1 | |||
| AY512927.1 | AAR87798.1 |
More Information
Set Membership: |
|

Genomic Map