Assay Details
Target Gene Details
Entrez Gene ID: | 14623 |
Gene Name: | gap junction protein, beta 6 |
Gene Aliases: |
AA958971, Cx30, D14Bwg0506e |
Location: |
Chr.14:57123301-57133611 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjb6 | NM_001010937.2 | 2 | 468 | NP_001010937.1 |
| NM_001271663.1 | 2 | 468 | NP_001258592.1 | |
| AK134617.1 | 2 | 467 | BAE22212.1 | |
| AK141610.1 | 2 | 468 | BAE24763.1 | |
| BC013811.1 | 2 | 135 | AAH13811.1 | |
| BC016507.1 | 2 | 143 | AAH16507.1 |
More Information
Set Membership: |
|

Genomic Map