Assay Details
Target Gene Details
Entrez Gene ID: | 239134 |
Gene Name: | guanylate cyclase 1, soluble, beta 2 |
Gene Aliases: |
B930025B03, GC-SB2 |
Location: |
Chr.14:62373023-62457562 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy1b2 | NM_001204340.1 | NP_001191269.1 | ||
| NM_172810.3 | NP_766398.1 | |||
| AK047129.1 | BAC32968.1 | |||
| AY236513.1 | AAO65585.1 | |||
| BC137868.1 | ||||
| BC144988.1 |
More Information
Set Membership: |
|

Genomic Map