Assay Details
Target Gene Details
Entrez Gene ID: | 268739 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 40 |
Gene Aliases: |
E130112L23Rik, Gm669, Solo |
Location: |
Chr.14:51984681-52006247 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef40 | NM_001145921.1 | 2 | 303 | NP_001139393.1 |
| NM_001145922.1 | 2 | 303 | NP_001139394.1 | |
| NM_198249.4 | 2 | 303 | NP_937892.2 | |
| AK053592.1 | 2 | 236 | ||
| AK131136.1 | 2 | 272 | BAD21386.1 | |
| AK134295.1 | 2 | 292 | BAE22088.1 | |
| AK143527.1 | 2 | 278 | BAE25418.1 | |
| AK147761.1 | 2 | 239 | BAE28120.1 | |
| AK155659.1 | 2 | 171 | BAE33372.1 | |
| AK163945.1 | 2 | 304 | ||
| AK202688.1 | ||||
| BC059018.1 | 2 | 205 | AAH59018.1 |
Target Gene Details
Entrez Gene ID: | 100502764 |
Gene Name: | predicted gene, 16617 |
Gene Aliases: |
- |
Location: |
Chr.14:51986389-51988829 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16617 | NR_045728.1 | |||
| AK136665.1 |
More Information
Set Membership: |
|

Genomic Map