Assay Details
Target Gene Details
Entrez Gene ID: | 13030 |
Gene Name: | cathepsin B |
Gene Aliases: |
CB |
Location: |
Chr.14:63122462-63145923 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsb | NM_007798.3 | NP_031824.1 | ||
| AK002456.1 | ||||
| AK083393.1 | BAC38900.1 | |||
| AK147192.1 | BAE27751.1 | |||
| AK149884.1 | BAE29144.1 | |||
| AK151790.1 | BAE30691.1 | |||
| AK153047.1 | ||||
| AK159992.1 | BAE35542.1 | |||
| AK167361.1 | BAE39458.1 | |||
| AK170959.1 | BAE42140.1 | |||
| AK190209.1 | ||||
| AK190539.1 | ||||
| AK191055.1 | ||||
| AK191391.1 | ||||
| AK196955.1 | ||||
| AK198213.1 | ||||
| AK199268.1 | ||||
| AK201893.1 | ||||
| AK202291.1 | ||||
| AK203639.1 | ||||
| BC006656.1 | AAH06656.1 | |||
| M14222.1 | AAA37494.1 | |||
| S69034.1 | AAB20536.1 |
More Information
Set Membership: |
|

Genomic Map