Assay Details
Target Gene Details
Entrez Gene ID: | 13030 |
Gene Name: | cathepsin B |
Gene Aliases: |
CB |
Location: |
Chr.14:63122462-63145923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 13030 |
Gene Name: | cathepsin B |
Gene Aliases: |
CB |
Location: |
Chr.14:63122462-63145923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Set Membership: |
|