Assay Details
Target Gene Details
Entrez Gene ID: | 13030 |
Gene Name: | cathepsin B |
Gene Aliases: |
CB |
Location: |
Chr.14:63122462-63145923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsb | NM_007798.3 | NP_031824.1 | ||
| AK002456.1 | ||||
| AK083393.1 | BAC38900.1 | |||
| AK147192.1 | BAE27751.1 | |||
| AK149884.1 | BAE29144.1 | |||
| AK151790.1 | BAE30691.1 | |||
| AK153047.1 | ||||
| AK159992.1 | BAE35542.1 | |||
| AK167361.1 | BAE39458.1 | |||
| AK170959.1 | BAE42140.1 | |||
| AK179071.1 | ||||
| AK190041.1 | ||||
| AK190750.1 | ||||
| AK191341.1 | ||||
| AK191739.1 | ||||
| AK191997.1 | ||||
| AK192159.1 | ||||
| AK192561.1 | ||||
| AK192818.1 | ||||
| AK194693.1 | ||||
| AK195254.1 | ||||
| AK196522.1 | ||||
| AK196531.1 | ||||
| AK197104.1 | ||||
| AK197767.1 | ||||
| AK197917.1 | ||||
| AK198000.1 | ||||
| AK198001.1 | ||||
| AK198619.1 | ||||
| AK198709.1 | ||||
| AK199167.1 | ||||
| AK200297.1 | ||||
| AK201305.1 | ||||
| AK201756.1 | ||||
| AK202640.1 | ||||
| AK203701.1 | ||||
| AK205609.1 | ||||
| AK209054.1 | ||||
| AK209352.1 | ||||
| AK210127.1 | ||||
| AK213062.1 | ||||
| AK213791.1 | ||||
| BC006656.1 | AAH06656.1 | |||
| M14222.1 | AAA37494.1 | |||
| S69034.1 | AAB20536.1 | |||
| Y18463.1 | CAA77181.1 |
More Information
Set Membership: |
|

Genomic Map