Assay Details
Target Gene Details
Entrez Gene ID: | 238988 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
6430531D06, CAST, CAST1/ERC2, D14Ertd171e, ELKS2alpha |
Location: |
Chr.14:27622410-28478537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erc2 | NM_177814.4 | NP_808482.2 | ||
| AK122265.1 | BAC65547.1 | |||
| AK147213.1 | BAE27770.1 | |||
| AY356530.1 | AAR14793.1 | |||
| AY356531.1 | AAR14794.1 | |||
| AY356532.1 | AAR14795.1 | |||
| AY356533.1 | AAR14796.1 | |||
| BC056760.1 | AAH56760.1 |
More Information
Set Membership: |
|

Genomic Map