Assay Details
Target Gene Details
Entrez Gene ID: | 13592 |
Gene Name: | early B cell factor 2 |
Gene Aliases: |
D14Ggc1e, EBF-2, Mmot1, O/E-3, OE-3 |
Location: |
Chr.14:67233292-67430952 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf2 | NM_001276387.1 | 16 | 2624 | NP_001263316.1 |
| NM_010095.6 | 16 | 2619 | NP_034225.1 | |
| AK053013.1 | 16 | 2620 | BAC35239.1 | |
| AK139262.1 | 16 | 2331 | BAE23936.1 | |
| BC049188.1 | 15 | 1918 | AAH49188.1 | |
| BC050922.1 | 15 | 2141 | AAH50922.1 | |
| U71189.1 | 15 | 1804 | AAB58423.1 | |
| U82441.1 | 15 | 1673 | AAC64322.1 | |
| U92703.1 | 15 | 2024 | AAB58323.1 |
More Information
Set Membership: |
|

Genomic Map