Assay Details
Target Gene Details
Entrez Gene ID: | 100503545 |
Gene Name: | nuclear GTPase, germinal center associated |
Gene Aliases: |
Gm600, SLIP-GC |
Location: |
Chr.14:65593499-65648444 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nuggc | NM_001195674.2 | NP_001182603.2 |

Genomic Map