Assay Details
Target Gene Details
Entrez Gene ID: | 68177 |
Gene Name: | emopamil binding protein-like |
Gene Aliases: |
5730442K12Rik, EBRP |
Location: |
Chr.14:61339763-61360445 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebpl | NM_026598.3 | NP_080874.2 | ||
| AF243434.2 | AAK28349.2 | |||
| AK011237.1 | BAB27485.1 | |||
| AK019140.1 | BAB31565.1 | |||
| AK019963.1 | BAB31938.1 | |||
| AK044618.1 | ||||
| AK163371.1 | BAE37321.1 | |||
| BC027422.1 | AAH27422.1 | |||
| BY349880.1 |
More Information
Set Membership: |
|

Genomic Map